missing translation for 'onlineSavingsMsg'
Learn More

ICI 192,605, Tocris Bioscience™

Potent thromboxane A2/TP receptor antagonist

£149.94 - £573.30

Chemical Identifiers

CAS 117621-64-4
Molecular Formula C22H23ClO5
Molecular Weight (g/mol) 402.871
InChI Key WHUIENZXNGAHQI-BNGKFCBFSA-N
Synonym d0b9na, 6-2beta-2-chlorophenyl-6beta-2-hydroxyphenyl-1,3-dioxan-5beta-yl-4-hexenoic acid, e-6-2s,4s,5r-2-2-chlorophenyl-4-2-hydroxyphenyl-1,3-dioxan-5-yl hex-4-enoic acid
PubChem CID 23654676
IUPAC Name (E)-6-[(2S,4S,5R)-2-(2-chlorophenyl)-4-(2-hydroxyphenyl)-1,3-dioxan-5-yl]hex-4-enoic acid
SMILES C1C(C(OC(O1)C2=CC=CC=C2Cl)C3=CC=CC=C3O)CC=CCCC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13438063 View Documents Tocris (Bio-Techne)
083710
10mg £149.94
10mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 18-12-2019
Please to purchase this item. Need a web account? Register with us today!
13448063 View Documents Tocris (Bio-Techne)
083750
50mg £573.30
50mg View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent thromboxane A2 receptor (TP receptor) antagonist. Competitively inhibits contractile responses to U 46619 (Cat. No. 1932).
Specifications

Chemical Identifiers

117621-64-4
402.871
d0b9na, 6-2beta-2-chlorophenyl-6beta-2-hydroxyphenyl-1,3-dioxan-5beta-yl-4-hexenoic acid, e-6-2s,4s,5r-2-2-chlorophenyl-4-2-hydroxyphenyl-1,3-dioxan-5-yl hex-4-enoic acid
(E)-6-[(2S,4S,5R)-2-(2-chlorophenyl)-4-(2-hydroxyphenyl)-1,3-dioxan-5-yl]hex-4-enoic acid
C22H23ClO5
WHUIENZXNGAHQI-BNGKFCBFSA-N
23654676
C1C(C(OC(O1)C2=CC=CC=C2Cl)C3=CC=CC=C3O)CC=CCCC(=O)O

Specifications

10mg
117621-64-4
d0b9na, 6-2beta-2-chlorophenyl-6beta-2-hydroxyphenyl-1,3-dioxan-5beta-yl-4-hexenoic acid, e-6-2s,4s,5r-2-2-chlorophenyl-4-2-hydroxyphenyl-1,3-dioxan-5-yl hex-4-enoic acid
C1C(C(OC(O1)C2=CC=CC=C2Cl)C3=CC=CC=C3O)CC=CCCC(=O)O
402.871
402.87
ICI 192, 605
C22H23ClO5
WHUIENZXNGAHQI-BNGKFCBFSA-N
(E)-6-[(2S,4S,5R)-2-(2-chlorophenyl)-4-(2-hydroxyphenyl)-1,3-dioxan-5-yl]hex-4-enoic acid
23654676
>98%