missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Icatibant, MedChemExpress
Description
Icatibant (HOE-140) is a potent and specific peptide antagonist of bradykinin B2 receptor with IC50 and Ki of 1.07 nM and 0.798 nM respectively.
Specifications
Specifications
| Chemical Name or Material | Icatibant |
| CAS | 130308-48-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C59H89N19O13S |
| Quantity | 10 mg |
| Synonym | HOE 140 |
| Solubility Information | H2O : 100 mg/mL (76.66 mM; Need ultrasonic) |
| SMILES | [H][C@]1(C[C@@H](C(N[C@H](C(O)=O)CCCNC(N)=N)=O)N2C([C@@H]3N(C([C@H](CO)NC([C@H](CC4=CC=CS4)NC(CNC([C@@H]5C[C@@H](O)CN5C([C@H]6N(C([C@H](CCCNC(N)=N)NC([C@H](N)CCCNC(N)=N)=O)=O)CCC6)=O)=O)=O)=O)=O)CC7=C(C=CC=C7)C3)=O)[C@]2([H])CCCC1 |
| Molecular Weight (g/mol) | 1304.52 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?