missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ICAM-5 Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | ICAM-5 |
---|---|
Clone | 283607 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Description
ICAM-5 Monoclonal specifically detects ICAM-5 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
ICAM-5 | |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q9UMF0 | |
7087 | |
Mouse myeloma cell line NS0-derived recombinant human ICAM-5 Ala28-Glu570 Accession # Q9UMF0 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ICAM-5 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) CD31, rhCEACAM-1, recombinant mouse (rm) DCC, rhICAM-1, -2, -3, rmICAM-5, rhMAdCAM-1, or rmVCAM-1 is observed. |
283607 | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ICAM5, ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, Telencephalin, TLCNTelencephalin, TLNtelencephalin | |
ICAM5 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only