Learn More
ICA 069673, Tocris Bioscience™
KV7.2/KV7.3 channel opener
Brand:  RD Systems (Bio-Techne) 4305/10
Code : Z1
Additional Details : Weight : 0.00010kg
Quantity 10mg
 View more versions of this product
Product Code. 13464524
  • £157.08 / 10mg
Estimated Shipment
05-04-2019
Add to basket
Description
Description
Selective KV7.2/KV7.3 (KCNQ2/3) channel opener (EC50 = 0.69μM). Exhibits 20-fold selectivity for KV7.2/KV7.3 over KV7.3/KV7.5, with no measurable activity over a panel of cardiac ion channels. Hyperpolarizes resting membrane potential and inhibits spontaneous action potentials in guinea pig detrusor muscle (DSM) isolated cells. Orally active in animal models of epilepsy.
Specifications
Chemical Identifiers
582323-16-8
269.636
n-2-chloropyrimidin-5-yl-3,4-difluorobenzamide, ica hplc, n-2-chloro-5-pyrimidinyl-3,4-difluorobenzamide, n-2-chloro-pyrimidin-5-yl-3,4-difluoro-benzamide
N-(2-chloropyrimidin-5-yl)-3,4-difluorobenzamide
C11H6ClF2N3O
IIBSHMFXVWTQSJ-UHFFFAOYSA-N
10149311
C1=CC(=C(C=C1C(=O)NC2=CN=C(N=C2)Cl)F)F
Specifications
ICA 069673
582323-16-8
n-2-chloropyrimidin-5-yl-3,4-difluorobenzamide, ica hplc, n-2-chloro-5-pyrimidinyl-3,4-difluorobenzamide, n-2-chloro-pyrimidin-5-yl-3,4-difluoro-benzamide
C1=CC(=C(C=C1C(=O)NC2=CN=C(N=C2)Cl)F)F
269.636
269.63
10mg
C11H6ClF2N3O
IIBSHMFXVWTQSJ-UHFFFAOYSA-N
N-(2-chloropyrimidin-5-yl)-3,4-difluorobenzamide
10149311
>99%
Videos