missing translation for 'onlineSavingsMsg'
Learn More

IBC 293, Tocris Bioscience™

Selective agonist for the GPR109B (HM74) receptor. Antilipolytic

Brand:  Tocris (Bio-Techne) 246950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461274

  • £336.61 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Highly selective agonist for GPR109B (HM74), human orphan G-protein-coupled receptor expressed in adipocytes. Selective for GPR109B over niacin receptor GPR109A (HM74A); EC50 values are 400 nM and > 1 μM respectively. Inhibits isoproterenol-stimulated lipolysis in vitro in isolated human adipocytes.
Specifications

Chemical Identifiers

306935-41-1
205.217
1-isopropyl-1h-1,2,3-benzotriazole-5-carboxylic acid, 1-isopropyl-1h-benzo d 1,2,3 triazole-5-carboxylic acid, 1-isopropyl-1,2,3-benzotriazole-5-carboxylic acid, 1-isopropyl-1h-benzotriazole-5-carboxylic acid, 1h-benzotriazole-5-carboxylic acid, 1-1-methylethyl, 1-isopropylbenzotriazole-5-carboxylic acid, 1h-benzotriazole-5-carboxylicacid, 1-1-methylethyl, 1-propan-2-yl-1h-1,2,3-benzotriazole-5-carboxylic acid, 1-methylethyl benzotriazole-5-carboxylic acid, maybridge1_005244
1-propan-2-ylbenzotriazole-5-carboxylic acid
C10H11N3O2
RUTVRAJKELSHCC-UHFFFAOYSA-N
2736690
CC(C)N1C2=C(C=C(C=C2)C(=O)O)N=N1

Specifications

IBC 293
306935-41-1
1-isopropyl-1h-1,2,3-benzotriazole-5-carboxylic acid, 1-isopropyl-1h-benzo d 1,2,3 triazole-5-carboxylic acid, 1-isopropyl-1,2,3-benzotriazole-5-carboxylic acid, 1-isopropyl-1h-benzotriazole-5-carboxylic acid, 1h-benzotriazole-5-carboxylic acid, 1-1-methylethyl, 1-isopropylbenzotriazole-5-carboxylic acid, 1h-benzotriazole-5-carboxylicacid, 1-1-methylethyl, 1-propan-2-yl-1h-1,2,3-benzotriazole-5-carboxylic acid, 1-methylethyl benzotriazole-5-carboxylic acid, maybridge1_005244
CC(C)N1C2=C(C=C(C=C2)C(=O)O)N=N1
205.217
205.22
50mg
C10H11N3O2
RUTVRAJKELSHCC-UHFFFAOYSA-N
1-propan-2-ylbenzotriazole-5-carboxylic acid
2736690
>99%
Videos