missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Nα-(3-Maleimidylpropionyl)Biocytin
Nα-(3-Maleimidylpropionyl)Biocytin
Brand: Molecular Probes™ M1602
Code : 50
Additional Details : Weight : 0.00525kg
Description
The thiol-reactive Na-(3-maleimidylpropionyl) biocytin can be used to attach biotin to biomolecules that can be subsequently detected with avidin or streptavidin conjugates.Specifications
25 mg | |
Nα-(3-Maleimidylpropionyl)Biocytin | |
Store in freezer -5°C to -30°C. | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.