Learn More
IkB zeta Monoclonal Antibody (LK2NAP), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 46-6801-82
184.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The LK2NAP monoclonal antibody reacts with mouse IkB zeta (INAP, MAIL). IkBz is a member of the IkB family of cytoplasmic proteins and transcription factors and also shares structural homology with Bcl-3. It is induced in monocytes, macrophages and B cells in response to TLR and IL-1R signaling. In NK cells, IkBz is rapidly upregulated in response to IL-12 and IL-18 and is necessary for production of IFN gamma. In T cells, IkBz is not necessary for IFN gamma and IL-4 production by Th1 and Th2 cells, respectively. However in Th17 cells, IkBz acts in concert with RORa and RORgt to upregulate IL-17A, IL-17F, IL-21, IL-22 and IL-23R expression. Applications Reported: This LK2NAP antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This LK2NAP antibody has been tested by intracellular staining and flow cytometric analysis of LPS-activated mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
IkB zeta Monoclonal antibody specifically detects IkB zeta in Mouse samples. It is validated for Flow Cytometry.Specifications
IkB zeta | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9EST8 | |
NFKBIZ | |
Affinity chromatography | |
RUO | |
80859 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
LK2NAP | |
PerCP-eFluor 710 | |
NFKBIZ | |
AA408868; FLJ30225; FLJ34463; Ikappa B-zeta variant 3; ikappaBzeta; I-kappa-B-zeta; IkappaB-zeta; IKBZ; ikB-zeta; IL-1 inducible nuclear ankyrin-repeat protein; INAP; Mail; molecule possessing ankyrin repeats induced by lipopolysaccharide; molecule possessing ankyrin-repeats induced by lipopolysaccharide; NF-kappa-B inhibitor zeta; NFKB inhibitor zeta; NFKBIZ; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, zeta; OTTHUMP00000214340; RGD1310834 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.