missing translation for 'onlineSavingsMsg'
Learn More

IkB zeta, clone: LK2NAP, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-6801-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15207957

  • £61.69 / 25µg


Estimated Shipment
26-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The LK2NAP monoclonal antibody reacts with mouse IkB zeta (INAP, MAIL). IkBz is a member of the IkB family of cytoplasmic proteins and transcription factors and also shares structural homology with Bcl-3. It is induced in monocytes, macrophages and B cells in response to TLR and IL-1R signaling. In NK cells, IkBz is rapidly upregulated in response to IL-12 and IL-18 and is necessary for production of IFN gamma. In T cells, IkBz is not necessary for IFN gamma and IL-4 production by Th1 and Th2 cells, respectively. However in Th17 cells, IkBz acts in concert with RORa and RORgt to upregulate IL-17A, IL-17F, IL-21, IL-22 and IL-23R expression.Applications Reported: This LK2NAP antibody has been reported for use in western blotting.Applications Tested: This LK2NAP antibody has been tested by western blotting on reduced lysates at > 1 ug/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

I kappa B zeta
LK2NAP
Unconjugated
Purified
Rat
25μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
NF-kappa-B inhibitor zeta, Nfkbiz
IgG2a
RUO
Primary
Murine
Videos