Learn More
Thermo Scientific™ Hypersil™ PREP HS Phenyl Bulk Media
Ensure optimal chromatography performance and cost savings with Hypersil PREP HS Phenyl bulk chromatography media.
Brand: Thermo Scientific™ 41510-10000B
Additional Details : CAS Number : 72245-35-3 Weight : 18.00000kg
Description
Ensure optimal chromatography performance with our Thermo Scientific™ Hypersil™ PREP HS Phenyl bulk chromatography media. Purchase bulk quantities of moderately hydrophobic, high surface area, phenyl-bonded media to repack preparative HPLC columns yourself without paying for new hardware.
Repack Columns Yourself with Bulk Media
Unique Selectivity
- Enhanced selectivity for aromatic and moderately polar analytes.
- Selectivity complementary to that provided by alkyl chain phases.
- Moderate hydrophobicity compared to C18.
- Phenyl (USP L11) selectivity - high surface area particle with 100Å pore size.
- pH stable from 2 to 8.
- Maximum temperature of 60°C.
Customer Specific Orders
- Please contact your local sales representative for Customer Specific Orders.
Specifications
10 μm | |
Phenyl (Ph) | |
10000 g | |
Prep LC | |
Yes | |
9% | |
L11 |
100 Å | |
2 to 8 | |
350 m²/g | |
Reversed Phase | |
Spherical, Fully Porous, Ultrapure Silica | |
Reversed Phase | |
60°C |