Thermo Scientific™ HyperSep™ Verify CX Plates
Get improved analysis of drugs of abuse, including basic and neutral drugs, with these SPE well plates featuring both nonpolar and ionic separation characteristics.
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: 60300-806
HYPERSEP-96 VERIFY-CX 25MG/1ML
UNSPSC: 41115712
Code: 16
Additional Details:
Additional Details: Weight: 0.10000kg
Product Code. 10424593
Quantity Price
1 £ 265.0 / Each
Estimated Shipment
20-02-2017
Add to basket
Description and Specification
Specification
Particle Size 40 - 60µm
For Use With (Equipment) Analysis of Drugs of Abuse
Type 96-well Plate
Description Verify-CX 25mg/1mL 96 Removable Well Plate, 1pk
Sorbent HyperSep Verify CX
Item Description Verify-CX 25mg/1mL 96 Removable Well Plate, 1pk
For Use With Analysis of Drugs of Abuse
The sorbent used in HyperSep Verify CX 96-Well Plates and Individual Wells is based on two functional groups: reversed phase C8 and a benzene sulfonic acid ion exchanger. The chemistry is co-polymerized on a rigid, purified silica gel support.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.