Learn More
Thermo Scientific™ HyperSep™ Universal Vacuum Well Extraction Plate and Plugs
Use these 24 or 48 well extraction plates and plugs with the Thermo Scientific™ HyperSep™ Universal Vacuum Manifold.
Brand: Thermo Scientific™ 60104-284
Code : NEW
Additional Details : Weight : 0.20000kg
Description
Use these 24 or 48 well extraction plates and plugs with Thermo Scientific™ HyperSep™ Universal Vacuum Manifold. This manifold process your samples in solid phase extraction SPE cartridges or 96 well plates.
Specifications
24 Well Extraction Plate, 1 pk | |
Universal Vacuum Manifold | |
24 Well Extraction Plate |
SPE Sample Processing | |
HyperSep |