missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ SAX Cartridges

Obtain excellent performance for extraction of carboxylic and other weak acids with these strong anion exchanger SPE cartridges.

Brand:  Thermo Scientific™ 60108-417

Code : 16

Additional Details : Weight : 2.26800kg

 View more versions of this product

Product Code. 10575615

  • £213.07 / Pack of 100
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Thermo Scientific™ HyperSep™ SAX Cartridges feature a strong anion exchange sorbent for extraction of weak acids. Typical applications include:

  • Isolation of anionic proteins
  • Removal of acidic food pigments
  • Isolation of phenolic compounds
  • Nucleic acids, nucleotides and surfactants

HyperSep SAX Strong Anion Exchanger Cartridges are ideally suited for the extraction of carboxylic and other weak acids. The chemistry extracts negatively charged compounds from both aqueous and nonaqueous solutions.

Amine group masks the effect of the carbon chain of the functional group

Selectivity can be tuned by choice of buffer

HyperSep SPE Cartridges and Plates

Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.

The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.

Specifications

Specifications

1 mL
50 mg
Thermo Scientific HyperSep-96 System
HyperSep
Solid Phase Extraction Cartridge
Pack of 100
50 mg Bed Weight; 1 mL column volume
Extraction of Weak Acids
40 to 60 μm
HyperSep SAX
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.