missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ Retain AX Cartridges

Achieve superior recoveries for acidic analytes with Thermo Scientific™ HyperSep™ Retain AX SPE Cartridges.

Brand:  Thermo Scientific™ 60107-417

Code : A0

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 12106590

  • £136.00 / Pack of 50


Estimated Shipment
31-05-2018
Add to basket

Specifications

Specifications

Bed weight 500mg/15mL Cartridge pk20
Recovery for Acidic Analyte
Extraction Column
500mg
Recovery for Acidic Analyte
HyperSep Retain AX
30 - 50µm
15mL
Videos

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.