Learn More
Thermo Scientific™ HyperSep™ Polystyrene Lab Plates
Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.
Brand:  Thermo Scientific™ 60110-208
UNSPSC : 41113036
Code : 16
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10524665
  • £311.00 / Pack of 5
Estimated Shipment
25-10-2017
Add to basket
Description
Description
The convenient 96-well HyperSep Lab Plates can be processed manually or using a liquid-handling robot. They are not suitable for use with a vacuum.
Specifications
Specifications
SCX; Polystyrene
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
96-well Plate
SCX; Polystyrene
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
HyperSep SCX
40 - 60┬Ám