Learn More
Thermo Scientific™ HyperSep™ Polypropylene Lab Plates
Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.
Brand:  Thermo Scientific™ 60110-308
Code : 16
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10714657
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
The convenient 96-well HyperSep Lab Plates can be processed manually or using a liquid-handling robot. They are not suitable for use with a vacuum.
Specifications
Specifications
SCX; Polypropylene
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
96-well Plate
SCX; Polypropylene
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
HyperSep SCX
40 - 60┬Ám
Videos
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.