Learn More
Thermo Scientific™ HyperSep™ Phenyl Plates
Obtain improved extraction of aromatic compounds and basic analytes with these phenyl SPE well plates offering alternative selectivity for retention of basic compounds.
Brand:  Thermo Scientific™ 60300-685
UNSPSC : 41115712
Code : 16
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10561105
  • £231.00 / Each
Estimated Shipment
25-07-2017
Add to basket
Specifications
Description
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Description & Specifications
Specifications
96-Well Plate; Bed weight 10mg
Extraction of Aromatic Compounds and Basic Analytes
96-well Plate
96-Well Plate; Bed weight 10mg
Extraction of Aromatic Compounds and Basic Analytes
HyperSep Phenyl
40 - 60µm