Learn More
Thermo Scientific™ HyperSep™ Florisil Plates
Isolate polar compounds from nonpolar matrices with these Florisil SPE well plates, which are especially well-suited for extraction of chlorinated pesticides.
Brand:  Thermo Scientific™ 60300-725
UNSPSC : 41115712
Code : 16
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10621134
  • £241.00 / Each
Estimated Shipment
23-08-2017
Add to basket
Specifications
Description
Extremely polar in nature, HyperSep Florisil SPE 96-Well Plates and Individual Wells are especially well-suited for extraction of chlorinated pesticides. The magnesia-loaded silica gel offers rapid flow rates for large sample volumes.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Description & Specifications
Specifications
Florisil 10mg/1mL 96 Removable Well Plate, 1pk
Isolation of Polar Compounds from Non-polar Matrices
96-well Plate
Florisil 10mg/1mL 96 Removable Well Plate, 1pk
Isolation of Polar Compounds from Non-polar Matrices
HyperSep Magnesia-loaded Silica Gel
40 - 60µm