Thermo Scientific™ HyperSep™ Filter Plates, 5-7¦L Bed Volume
Get simple, effective clean-up of proteins, peptides, DNA, RNA and other biomolecules in small-scale samples with these 96-well filter plates.
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: 60110-405
UNSPSC: 41104923
Code: 16
Additional Details:
Additional Details: Weight: 0.30000kg
Disclaimers: Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Code. 10066174
Quantity Price
1 £ 272.0 / Each
Estimated Shipment
25-05-2017
Add to basket
Description and Specification
Specification
For Use With (Equipment) Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Description C18 plus Hypercarb; 5-7uL bed volume
Type 96-well Filter Plate
Sorbent Hypercarb and HyperSep C18 (Mix Mode)
Item Description C18 plus Hypercarb; 5-7uL bed volume
For Use With Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.