Learn More
Thermo Scientific™ HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE products for QuEChERS clean-up.
Brand:  Thermo Scientific™ 60105-214-P
Additional Details : Weight : 0.15300kg
 View more versions of this product
Product Code. 16883140
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Thermo Scientific™ HyperSep Dispersive SPE Clean-Up Products are available in a range of pre-prepared sorbent combinations, each containing the proper sorbents for optimum clean-up of analytes extracted using QuEChERS methods.
Comprehensive range of Dispersive SPE products that contain the proper sorbents foroptimum extraction, clean-up and separation of analytes from complex matrices such as food using QuEChERS methods.
  • Save valuable time in preparation – products are supplied pre-prepared
  • Reduced variability due to consistency in products and rigorous quality control
  • Cleaner extracts from cleaner, pre-prepared products
  • Lower peak counts as a result of using pre-prepared products
Specifications
Specifications
900 mg anhydrous magnesium sulfate, 300 mg PSA
Quechers
15 mL
QuEChERS Method
900 mg MgSO4, 300 mg PSA
50
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.