missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ C8 Plates

Need a less retentive alternative to C18 for polar and nonpolar compounds? These hydrophobic reversed phase C8 well plates offer excellent performance.

Brand:  Thermo Scientific™ 60307-211

Code : A0

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13466986

  • £249.36 / Each


Estimated Shipment
02-11-2020
Add to basket

Description

Description

HyperSep C8 SPE Well Plates and Individual Wells provide excellent performance characteristics for moderately polar analytes. They also offer the potential for polar interactions greater than with C18.

HyperSep SPE Cartridges and Plates

HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.

The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.

Specifications

Specifications

Screening of Drugs, Metabolites, and Peptides in Biological Samples
Solid Phase Extraction Well Plate
40 to 60 µm
10 mg
HyperSep
HyperSep C8
1 mL
96-Well Plate; Bed weight 10 mg/2 mL
1 Pack
Videos
Documents