Learn More
Ansell Edmont™ HyFlex™ 11-638 Series Gray Polyurethane Medium Weight Gloves
Yarn structure contains an optimal rate of HPPE. Offers high levels of cut resistance for a safe use in controlling, handling and assembly applications involving contacts with sharp pieces.
Brand:  Ansell Edmont™ 11638060
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12660752
  • £189.87 / 12 pairs
Estimated Shipment
11-06-2018
Add to basket
Description
Description
  • The Lycra™ in the liners of the gloves as well as the polyurethane coating provides excellent flexibility and fit.
  • The specific formulation of the polyurethane dipping provides a secure grip on dry or slightly oily pieces.
  • The PU formulation also increases abrasion resistance on the gloves’ work surface.
  • With strong cut resistance
  • Knitted construction
Specifications
Specifications
Polyurethane
Long knit wrist
Coated-palm
High cut resistance
144 Pairs
Medium Weight Gloves
Polyurethane
Nylon/Lycra®/Dyneema®/Cotton/Kevlar®
Kevlar®
6
CE Mark, Standard CATEGORY II EN 388 : 4443
Videos