missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Hydroxypropyl methylcellulose, 28-30% methoxyl, 7-12% hydroxypropyl, viscosity (2% aq. soln., 20°C) 7500-14000 mPa.s

Brand:  Alfa Aesar™ 045847.22

Code : NEW

Additional Details : CAS Number : 9004-65-3 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15405005

  • £22.00 / 100g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

Hydroxypropyl methylcellulose
28 to 30% methoxyl, 7 to 12% hydroxypropyl, viscosity (2% aq. soln., 20°C) 7500 to 14000mPa.s
14,4842
9004-65-3
MFCD00131360
Methocel; HPMC

Specifications

Hydroxypropyl methylcellulose
28 to 30% methoxyl, 7 to 12% hydroxypropyl, viscosity (2% aq. soln., 20°C) 7500 to 14000mPa.s
Powder
14,4842
100g
9004-65-3
MFCD00131360
Methocel; HPMC
Videos
SDS