missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.0 |
CAS Max % | 100.0 |
Density | 1.0300g/mL |
Boiling Point | 240.0°C |
Flash Point | 101°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% min. (Mixture of isomers) (GC) |
Packaging | Glass bottle |
Linear Formula | CH2=C(CH3)CO2CH2CH(OH)CH3 |
Refractive Index | 1.4470 to 1.4490 |
Show More |
Safety and Handling
Product Identifier
- 2-Hydroxypropyl methacrylate, mixture of isomers, stabilized
- Warning
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'