missing translation for 'onlineSavingsMsg'
Learn More

Hydroxymethylferrocene, 97%, Thermo Scientific™

Catalog Number p-7050672
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1273-86-5
C11H12FeO
216.06
MFCD00040878
KLMBXXGIFRROFL-UHFFFAOYSA-N
Ferrocenemethanol
[Fe].c1cccc1.OCc1cccc1
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1273-86-5
C11H12FeO
216.06
MFCD00040878
KLMBXXGIFRROFL-UHFFFAOYSA-N
Ferrocenemethanol
[Fe].c1cccc1.OCc1cccc1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 79°C to 81°C
Color Yellow
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information It is Soluble in water (partly miscible),and methanol.
Formula Weight 216.07
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Hydroxymethylferrocene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'