missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Hydroxymethylferrocene, 97%

Brand:  Alfa Aesar™ 039257.14

Code : NEW

Additional Details : CAS Number : 1273-86-5 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11323658

  • £959.70 / 25g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

Hydroxymethylferrocene
216.07
Ferrocenemethanol
1273-86-5
MFCD00040878

Specifications

Hydroxymethylferrocene
Yellow
25g
Crystalline
216.07
Ferrocenemethanol
Moisture Sensitive
79° to 81°C
1273-86-5
97%
MFCD00040878
Videos
SDS