missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylammonium Chloride, 98+%, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical H/1900/53

Code : 21

Additional Details : CAS Number : 5470-11-1 Weight : 0.50000kg Transport : UN number : 2923 Chem class : 8 Pack group : III


Quantity 500g

 View more versions of this product

Product Code. 10101860

  • £127.94 / 500g

In stock, ready to ship: 48

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

5470-11-1
69.49
WTDHULULXKLSOZ-UHFFFAOYSA-N
443297
hydroxylamine;hydrochloride
ClH4NO
51089
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
CHEBI:5807
NO.Cl

Specifications

5470-11-1
51089
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydroxylamine;hydrochloride
443297
69.49g/mol
Plastic powder jar
155°C
500g
ClH4NO
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
NO.Cl
69.49
CHEBI:5807
Solid
White
2.5 to 3.5
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydroxylamine hydrochloride

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.

SDS Documents
Certificates