missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyguanidine sulfate, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mg
Description
Reacts with nitric oxide forming an adduct which is a potent and stable vasodilator
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 6345-29-5 |
Melting Point | 132°C to 134°C (decomposition) |
Color | White |
Molecular Formula | CH5N3O·0.5H2SO4 |
MDL Number | MFCD00042033 |
Quantity | 50 mg |
Formula Weight | 124.11 |
Physical Form | Crystals |
Chemical Name or Material | Hydroxyguanidine sulfate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'