missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone, 99%, Thermo Scientific Chemicals

Catalog Number 10500391
24.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
123-31-9
C6H6O2
110.11
MFCD00002339
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
785
CHEBI:17594
benzene-1,4-diol
C1=CC(=CC=C1O)O
123-31-9
C6H6O2
110.11
MFCD00002339
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
785
CHEBI:17594
benzene-1,4-diol
C1=CC(=CC=C1O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 170.0°C to 174.0°C
Color White
Density 1.32
Boiling Point 285.0°C to 287.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 06, 836
Fieser 05,341; 14,249
Merck Index 15, 4845
Solubility Information Solubility in water: 70g/L in water (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene,readily soluble in ethanol,acetone and methanol
Formula Weight 110.11
Percent Purity 99%
Physical Form Needle-like Crystals or Crystalline Powder
Chemical Name or Material Hydroquinone, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydroquinone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'