Learn More
Contec™ HydroPure
Contec HydroPure is a blend of 6% hydrogen peroxide and purified water.
Brand: Contec™ FBC16HP
Additional Details : Weight : 6.89000kg
Description
Efficacious against bacteria, fungi, moulds, yeasts and spores, Contec HydroPure leaves little to no residue and is ideal for use on most surfaces. Contec HydroPure is available in a 1 L trigger spray and 1 L capped container as well as a 5 L capped container. A sterile version for more critical areas is also available. The product is 0.2 μm filtered and filled under Grade A uni-directional flow air and bagged in a Grade B cleanroom.
Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms.
Features & Benefits
- Hydrogen peroxide breaks down to water and oxygen making it very low residue
- Filtered to 0.2 microns under Grade A airflow in a Grade B cleanroom ensuring the product is free from contamination and particulates
- Sporicidal in 15 mins
- Trigger spray and 'bag in bottle' protected system prevents bottle contents from becoming contaminated during use
- Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
- Double bagged in linear tear packaging so each bag is easy to open even when wearing gloves
- Not classed as corrosive and can be used safely in all areas of the cleanroom
Specifications
6% hydrogen peroxide and purified water | |
Capped Bottle | |
1 L |
Safety and Handling
- CONTEC STERILE & FILTERED CAPPED HYDROPURE 1L
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : CONTEC STERILE & FILTERED CAPPED HYDROPURE 1L