Learn More
Honeywell™ HYDROGRAIN Gloves
Honeywell™ HYDROGRAIN Gloves designed for handling indoor and outdoor operations in a damp or greasy environment.
Brand:  Honeywell™ 2049260-08
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12651103
  • £13.86 / Each
Estimated Shipment
15-04-2021
Add to basket
Description
Description
  • 5 finger leather glove enriched by a special tanning process to make the glove more water repellent
  • Gunn cut wrist type
  • Winged thumb makes the glove more anatomical for greater ease of use
  • Fully water-repellent grain cowhide leather
  • Elasticated wrist offers added support
  • Outstanding comfort and flexibility
Specifications
Specifications
Cowhide Leather
Yellow
EN 420-2003 EEC Basic standards, EN 388-2016 Mechanical hazards
8
25 cm