missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide USP, Macron Fine Chemicals™

macron

Brand:  Macron Fine Chemicals 5232-10

Additional Details : Weight : 4.00000kg

Packaging Glass
Quantity 4L

 View more versions of this product

Product Code. 15578324

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

7722-84-1
34.014
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
CHEBI:16240
OO
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
hydrogen peroxide

Specifications

Hydrogen Peroxide
7722-84-1
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
OO
34.014
CHEBI:16240
USP
4L
Topical Solution
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
784
34.01
Glass
Videos
Documents