missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, Stabilized, 0.3, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5383299
Click to view available options
Quantity:
500 mL
Packaging:
HDPE Pressure Bottom Poly Bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7722-84-1
H2O2
34.014
MHAJPDPJQMAIIY-UHFFFAOYSA-N
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
784
CHEBI:16240
hydrogen peroxide
OO
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7722-84-1
H2O2
34.014
MHAJPDPJQMAIIY-UHFFFAOYSA-N
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
784
CHEBI:16240
hydrogen peroxide
OO

High quality chemicals for laboratory and specialized industrial use

Quantity 500 mL
Formula Weight 34.01
Percent Purity 30%
Grade Meets ACS reagent chemical requirements
Packaging HDPE Pressure Bottom Poly Bottle
Specific Gravity 1L = 1.11kg
Chemical Name or Material Hydrogen Peroxide, Stabilized
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'