missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide, ACS reagent, 30 wt% solution in water, nonstabilized, ACROS Organics™

Brand:  Acros Organics 411880010

Code : 29

Additional Details : CAS Number : 7722-84-1 Weight : 1.05000kg Transport : UN number : 2014 Chem class : 2 Pack group : II


Quantity 1kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10318293

  • £41.70 / 1kg


In Stock 1
EU Stock 51
Add to basket


Specifications

Chemical Identifiers

Hydrogen peroxide
ACS Reagent Grade
H2O2
34
7732-18-5
29.0 to 32.0%
HOOH
MFCD00011333

Specifications

Hydrogen peroxide
Vapor Pressure: 23mmHg at 30°C
ACS Reagent Grade
29.0 to 32.0%
H2O2
34
MFCD00011333
15, 4835
1ppm max.
2ppm max.
2ppm max.
Solubility in water: soluble. Other solubilities: soluble in ether, alcohol, insoluble in petroleum ether, decomposed by many organic solvents
5ppm max.
>125°C
Clear
1kg
7732-18-5
5ppm max.
Solution
3ppm max.
HOOH
1.1100g/mL
01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145
Solubility in water: soluble. Other solubilities: soluble in ether, alcohol, insoluble in petroleum ether, decomposed by many organic solvents
0.5ppm max.
Plastic bottle
0.002% max.
1.11
0.0006 meq/g max.
108.0°C
− 33.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydrogen peroxide 20-35wt% solution in water

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS