missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide 6% (w/w) (20 Volumes), Extra Pure SLR, Fisher Chemical™

Brand:  Fisher Chemical H/1600/15

Code : 22

Additional Details : CAS Number : 7722-84-1 Weight : 1.11296kg


Quantity 1L

 View more versions of this product

Product Code. 10020160

Quantity Price
1 £50.33 / 1 L
4 £42.72 / 1 L
16 £40.59 / 1 L
In stock, ready to ship: 39

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Chemical Identifiers

7722-84-1
34.014
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
hydrogen peroxide
H2O2
11333
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
CHEBI:16240
OO

Specifications

7722-84-1
11333
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
784
34.0128g/mol
HDPE plastic bottle
Colorless
2.5 to 3.5
H2O2
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
OO
34.014
CHEBI:16240
Liquid
100°C
0°C
1L
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydrogen peroxide 20 volumes >6% w/v

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Hydrogen peroxide

SDS Documents