missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number p-4228711
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
26023-84-7
Cl6H2Pt
409.80
MFCD00149909
ZKOQTCCVSRSECD-UHFFFAOYSA-J
Hexachloroplatinic acid hydrate, Platinic chloride hydrate
dihydrogen hexachloroplatinumtetrakis(ylium)
[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl
This item is not returnable. View return policy
26023-84-7
Cl6H2Pt
409.80
MFCD00149909
ZKOQTCCVSRSECD-UHFFFAOYSA-J
Hexachloroplatinic acid hydrate, Platinic chloride hydrate
dihydrogen hexachloroplatinumtetrakis(ylium)
[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 60.0°C
Physical Form Powder, Crystals and/or Chunks
Quantity 1 g
Assay Percent Range 37% min. (Pt)
Linear Formula H2PtCl6·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 409.82
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Hydrogen hexachloroplatinate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'