missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen chloride, nominally 2.5M in ethanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
Description
Hydrogen chloride, 2.5M. in ethanol is used in the manufacturing of pharmaceuticals to get a hydrochloride salt. For example, ranitidine base is converted to ranitidine hydrochloride, which is used as anti ulcerative drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
UN Number | UN2924 |
Concentration | Nominally 2.5M in ethanol |
Solubility Information | Miscible with water at room temperature. Also miscible with methanol,ethanol and ether. |
IUPAC Name | chlorane |
Formula Weight | 36.46 |
Quantity | 250 mL |
Physical Form | Liquid |
Chemical Name or Material | Hydrochloric acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'