Learn More
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 450720010
25.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 7647-01-0 Weight : 0.90900kg Transport : UN number : 2920 Chem class : 8 Pack group : I
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hydrochloric acid | |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.45 | |
Glass bottle | |
1 L |
7647-01-0,67-63-0 | |
HCl | |
0.9090g/mL | |
Solubility in water: >1000g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol: >50g/L | |
Cl | |
36.46 | |
CHEBI:17883 | |
Technical | |
0.909 | |
11°C |
Safety and Handling
- Hydrogen chloride, 4 to 6N solution in 2-propanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Isopropyl alcohol, Hydrogen chloride
RUO – Research Use Only