Learn More
hydrogen chloride, 1M solution in acetic acid, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 370271000
26.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 29
Additional Details : CAS Number : 7647-01-0 Weight : 0.10920kg Transport : UN number : 2920 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hydrochloric acid | |
1M solution in acetic acid | |
1M, exact strength on the certificate of analysis | |
AcroSeal™ Glass Bottle | |
40°C | |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 |
Clear colorless | |
7647-01-0,64-19-7 | |
1.0920g/mL | |
1.092 | |
100 mL | |
HCl | |
14, 4794 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.45 |
Safety and Handling
- Hydrogen chloride, 1.0M solution in Acetic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only