missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid, Thermo Scientific Chemicals

Catalog Number p-3776505
51.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
10035-10-6, 64-19-7
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
Br
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
10035-10-6, 64-19-7
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
Br

Hydrogen bromide, 31 to 34 wt% (HBr), BrH, CAS Number-10035-10-6, 64-19-7, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, Vapor Pressure: 300mmHg at 20 deg.C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Color Yellow
Physical Form Fuming Liquid
Quantity 500 mL
Linear Formula HBr
UN Number 2922
Fieser 01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273
Merck Index 09, 4665
IUPAC Name hydrogen bromide
Formula Weight 80.9
Percent Purity 31 to 34 wt% (HBr)
Grade Pure
Packaging Glass Bottle
Vapor Pressure 300mmHg at 20°C
Density 1.42 g/mL
Chemical Name or Material Hydrogen bromide, Pure
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrogen bromide, 33 wt% solution in glacial acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Acetic acid, Hydrogen bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'