missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid 47-51%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™

Catalog Number 11934101
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
PFA Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7664-39-3
FH
20.01
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
14917
CHEBI:29228
hydrogen fluoride
F
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7664-39-3
FH
20.01
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
14917
CHEBI:29228
hydrogen fluoride
F
pH 1
Color Colorless
Melting Point -35°C
Boiling Point 105°C
Packaging PFA Bottle
Certificate of Analysis Certificate of Lot included with each bottle.
Quantity 1 L
Physical Form Liquid
UN Number 1790
Merck Index 15, 4827
Formula Weight 20.01
Grade OPTIMA™
Chemical Name or Material Hydrofluoric Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrofluoric acid 47-51%, Optima for Ultra traces analysis
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen fluoride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'