missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, ACS, 48-51%, Thermo Scientific Chemicals

Catalog Number p-4408623
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7664-39-3
FH
20.01
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
14917
CHEBI:29228
fluorane
F
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7664-39-3
FH
20.01
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
14917
CHEBI:29228
fluorane
F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Appearance Colorless liquid
Color Colorless
Odor Acrid
Assay Percent Range 48-51%
Quantity 10 g
Physical Form Liquid
Formula Weight 20.01
Grade ACS
Chemical Name or Material Hydrofluoric acid, Special handling precautions required

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'