Learn More
Hydrofluoric acid, 99.99% (metals basis) 40% min, Thermo Scientific Chemicals
Description
Hydrofluoric acid is used in the glass etching, electronic and chemical industries. Hydrogen fluoride is mainly used in the production of organofluorine compounds, aluminum and chlorofluorocarbons. Hydrogen fluoride is also used for separating uranium isotopes, as a catalyst in the petroleum industry, and in stainless steel pickling. Hydrofluoric acid is used as a pickling agent and used in the semiconductor materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Boiling Point | 112°C |
Density | 1.16 g/mL |
Odor | Acrid |
Sensitivity | Hygroscopic |
Quantity | 1000 mL |
Physical Form | Liquid |
UN Number | UN1790 |
Merck Index | 14,4790 |
Solubility Information | Strongly miscible with water and alcohol. Slightly miscible with ether and miscible with many organic solvents. |
Show More |
RUO – Research Use Only