missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, 99.99% (metals basis) 40% min, Thermo Scientific Chemicals

Catalog Number 11321588
Click to view available options
Quantity:
25 mL
250 mL
1000 mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7664-39-3
FH
20.01
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
14917
CHEBI:29228
hydrogen fluoride
F
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7664-39-3
FH
20.01
MFCD00011346
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
14917
CHEBI:29228
hydrogen fluoride
F

Hydrofluoric acid is used in the glass etching, electronic and chemical industries. Hydrogen fluoride is mainly used in the production of organofluorine compounds, aluminum and chlorofluorocarbons. Hydrogen fluoride is also used for separating uranium isotopes, as a catalyst in the petroleum industry, and in stainless steel pickling. Hydrofluoric acid is used as a pickling agent and used in the semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Boiling Point 112°C
Density 1.16 g/mL
Odor Acrid
Sensitivity Hygroscopic
Quantity 250 mL
Physical Form Liquid
UN Number UN1790
Merck Index 14,4790
Solubility Information Strongly miscible with water and alcohol. Slightly miscible with ether and miscible with many organic solvents.
Formula Weight 20.01
Percent Purity 99.99%
Assay (metals basis) 40% min
Chemical Name or Material Hydrofluoric acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'