missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, TraceSELECT™ Ultra, Honeywell Fluka™

Catalog Number p-7072288
Click to view available options
Quantity:
250 mL
500 mL
1 L
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
hydrogen chloride
Cl
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
hydrogen chloride
Cl

TraceSELECT™ Ultra, for ultratrace analysis, 32-35%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY
Color Yellow
Concentration 32 to 35% (T)
Density 1.2g/mL at 25°C, 1.15g/mL at 20°C
Vapor Pressure 3.23 psi (21.1°C), 7.93 psi (37.7°C)
Quantity 250 mL
UN Number UN1789
Beilstein 1098214
Formula Weight 36.46g/mol
Grade TraceSELECT™ Ultra
Chemical Name or Material Hydrochloric acid
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'