missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid NF, Macron Fine Chemicals™
62.32 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
6 x 500 mL
Packaging:
Glass
Specifications
CAS | 7647-01-0 |
Packaging | Glass |
Quantity | 6 x 500 mL |
Molecular Formula | ClH |
MDL Number | MFCD00011324 MFCD00792839 |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
SMILES | Cl |
IUPAC Name | chlorane |
Molecular Weight (g/mol) | 36.46 |
Show More |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'