missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Hydrochloric acid, Environmental Grade Plus, 33-36%

Brand:  Alfa Aesar™ 038744.AP

Code : NEW

Additional Details : CAS Number : 7647-01-0 Weight : 0.01000kg Transport : UN number : 1789 Chem class : 8 Pack group : II

Product Code. 11480181

  • £504.00 / 500mL


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Specifications

Hydrochloric acid
7647-01-0
Liquid, Double Sub-Boiling Quartz Distillation
36.46
UN1789
1.19g/mL
81.5° to 110°C
500mL
Environmental Grade Plus
33 to 36%
MFCD00011324
14,4780
Pungent
Videos
SDS