missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, DILUT-IT Concentrate, 0.01M, J.T. Baker™
Click to view available options
Quantity:
For dilution to 1 L
Packaging:
LDPE ampule
Description
DILUT-IT solutions are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications. Simply dilute the volumetric concentrate to 1 liter (carefully following instructions provided with the package) and the resulting solution will have an exact normality within ±0.1%.
- Enhanced productivity: Eliminate expensive, time-consuming steps, such as the selection and analysis of starting reagents, and weighing and standardization of the final solution
- Space saving: Products require only one-tenth of the space required for ready-made solutions
- Reduced shipping costs: Spend less on shipping due to the reduced weight and package size of a concentrate
Specifications
Color | Colorless |
CAS | 7647-01-0 |
Melting Point | 0°C |
Concentration | 0.01mol/L |
Density | 1.02g/cm3 at 25°C |
Packaging | LDPE ampule |
Vapor Pressure | 1.100 hPa (50°C) |
Quantity | For dilution to 1 L |
Molecular Formula | ClH |
Physical Form | Liquid |
Show More |
Safety and Handling
Signal Word
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'