missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, ACS, HCl 36.5-38.0%, Thermo Scientific Chemicals

Catalog Number p-4781932
Click to view available options
Quantity:
6 x 2724 g
This item is not returnable. View return policy
7647-01-0
ClH
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
hydrogen chloride
Cl
This item is not returnable. View return policy
7647-01-0
ClH
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
hydrogen chloride
Cl

 

Acidizing petroleum wells, chemical intermediate, ore reduction, food processing, pickling and metal cleaning, general cleaning, and laboratory reagent.Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY
Appearance Colorless pungent
Color Colorless
Boiling Point 81.5°C to 110°C
Density 1.19
Odor Pungent
Assay Percent Range 36.5 to 38.0%
Quantity 6 x 2724 g
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride
Solubility Information Soluble aq. soln.,alcohol,ether and benzene.Completely miscible with water.
Formula Weight 36.46
Grade ACS
Chemical Name or Material Hydrochloric acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'