missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 99.99% Trace Metals Basis, Honeywell™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
pH | >0.1 (at 20°C) |
Color | Colorless |
Melting Point | -35°C |
Boiling Point | 42°C |
Density | 1.2g/mL at 25°C |
Odor | Stinging |
Packaging | Poly Bottle |
Vapor Pressure | 3.23psi (21.1°C), 7.93psi (37.7°C) |
Quantity | 500 mL |
Physical Form | Liquid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'