missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 6.0N Standardized Solution, Alfa Aesar™

Brand:  Alfa Aesar 044921.K7

Code : NEW

Additional Details : Weight : 4.00000kg Transport : UN number : 1789 Chem class : 8 Pack group : II

Quantity 4L

 View more versions of this product

Product Code. 15412077

  • £64.70 / 4 L


Estimated Shipment date
from Supplier 18-07-2018
Add to basket

Specifications

Chemical Identifiers

Hydrochloric acid
Liquid
36.46
7732-18-5
HCl
MFCD00011324

Specifications

Hydrochloric acid
Colorless to pale yellow liquid
HCl
MFCD00011324
23 hPa (17mm Hg) at 20°C
4L
7732-18-5
Liquid
36.46
Hydrochloric acid: 49.6%; Water: 50.4%
Colorless to pale yellow
Videos
SDS